Merck
  • Ethanol induces interferon expression in neurons via TRAIL: role of astrocyte-to-neuron signaling.
Ethanol induces interferon expression in neurons via TRAIL: role of astrocyte-to-neuron signaling.
Psychopharmacology (2019-01-06)
Colleen J Lawrimore, Leon G Coleman, Fulton T Crews
ABSTRACT
Alcohol use disorder (AUD) involves dysregulation of innate immune signaling in brain. Toll-like receptor 3 (TLR3), an innate immune receptor that is upregulated in post-mortem human alcoholics, leads to induction of interferon (IFN) signaling. IFNs have been linked to depressive-like symptoms and therefore may play a role in addiction pathology. Astrocyte-neuronal signaling may contribute to maladaptation of neuronal circuits. In this manuscript, we examine ethanol (EtOH) induction of IFN signaling in neuronal, astrocyte, and microglial cell lines and assess astrocyte-neuronal interactions. U373 astrocytes, SH-SY5Y neurons, and BV2 microglia were treated with EtOH and analyzed for autocrine/paracrine IFN signaling. EtOH induced TLR3, IFNβ, and IFNγ in SH-SY5Y neurons and U373 astrocytes, but not in BV2 microglia. The IFN response gene TRAIL was also strongly upregulated by TLR3 agonist Poly(I:C) and EtOH in U373 astrocytes. TRAIL blockage via neutralizing antibody prevented induction of IFNs in SH-SY5Y neurons but not in U373 astrocytes. Blocking TRAIL in conditioned media from EtOH-treated astrocytes prevented induction of IFNs in SH-SY5Y neurons. Finally, an in vivo model of chronic 10-day binge EtOH exposure in C57BL6/J mice, as well as single acute treatment with Poly(I:C), showed increased TRAIL +IR cells in both orbitofrontal and entorhinal cortex. This study establishes a role of astrocyte to neuron TRAIL release in EtOH-induced IFN responses. This may contribute to alcohol associated negative affect and suggest potential therapeutic benefit of TRAIL inhibition in AUD.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
TRAIL Protein, Recombinant human, Human TRAIL (TNF-Related Apoptosis Inducing Ligand), also called APO2 Ligand, is a cytotoxic protein which activates rapid apoptosis in tumor cells, but not in normal cells.