Merck
  • Synthesis of Site-Specific Radiolabeled Antibodies for Radioimmunotherapy via Genetic Code Expansion.
Synthesis of Site-Specific Radiolabeled Antibodies for Radioimmunotherapy via Genetic Code Expansion.
Bioconjugate chemistry (2016-10-21)
Yiming Wu, Hua Zhu, Bo Zhang, Fei Liu, Jingxian Chen, Yufei Wang, Yan Wang, Ziwei Zhang, Ling Wu, Longlong Si, Huan Xu, Tianzhuo Yao, Sulong Xiao, Qing Xia, Lihe Zhang, Zhi Yang, Demin Zhou
ABSTRACT
Radioimmunotherapy (RIT) delivers radioisotopes to antigen-expressing cells via monoantibodies for the imaging of lesions or medical therapy. The chelates are typically conjugated to the antibody through cysteine or lysine residues, resulting in heterogeneous chelate-to-antibody ratios and various conjugation sites. To overcome this heterogeneity, we have developed an approach for site-specific radiolabeling of antibodies by combination of genetic code expansion and click chemistry. As a proof-of-concept study, model systems including anti-CD20 antibody rituximab, positron-emitting isotope 64Cu, and a newly synthesized bifunctional linker (4-dibenzocyclooctynol-1,4,7,10-tetraazacyclotetradecane-1,4,7,10-tetraacetic acid, DIBO-DOTA) were used. The approach consists of three steps: (1) site-specific incorporation of an azido group-bearing amino acid (NEAK) via the genetic code expansion technique at the defined sites of the antibody as a "chemical handle"; (2) site-specific and quantitative conjugation of bifunctional linkers with the antibodies under a mild condition; and (3) radiolabeling of the chelate-modified antibodies with the appropriate isotope. We used heavy-chain A122NEAK rituximab as proof-of-concept and obtained a homogeneous radioconjugate with precisely two chelates per antibody, incorporated only at the chosen sites. The conjugation did not alter the binding and pharmacokinetics of the rituximab, as indicated by in vitro assays and in vivo PET imaging. We believe our research is a good supplement to the genetic code expansion technique for the development of novel radioimmunoconjugates.
Sigma-Aldrich
N6-((2-Azidoethoxy)carbonyl)-L-lysine hydrochloride, ≥95%