Merck
  • Rotenone and 3-bromopyruvate toxicity impacts electrical and structural cardiac remodeling in rats.
Rotenone and 3-bromopyruvate toxicity impacts electrical and structural cardiac remodeling in rats.
Toxicology letters (2019-10-05)
Chengchuang Zhan, Guangzhong Liu, Jianqiang Li, Guangnan Li, Tiankai Li, Hongyan Zhao, Luyifei Li, Wen Yang, Nan Bai, Min Zheng, Junfen Yang, Weimin Li
ABSTRACT
3-Bromopyruvate (3-BrPA) is a promising agent that has been widely studied in the treatment of cancer and pulmonary hypertension. Rotenone is a pesticide commonly used on farms and was shown to have anti-cancer activity and delay fibrosis progression in chronic kidney disease in a recent study. However, there are few studies showing the toxicity of rotenone and 3-BrPA in the myocardium. To support further medical exploration, it is necessary to clarify the side effects of these compounds on the heart. This study was designed to examine the cardiotoxicity of 3-BrPA and rotenone by investigating electrical and structural cardiac remodeling in rats. Forty male rats were divided into 4 groups (n = 10 in each group) and injected intraperitoneally with 3-BrPA, rotenone or a combination of 3-BrPA and rotenone. The ventricular effective refractory period (VERP), corrected QT interval (QTc), and ventricular tachycardia/ventricular fibrillation (VT/VF) inducibility were measured. The expression of Cx43, Kir2.1, Kir6.2, DHPRα1, KCNH2, caspase3, caspase9, Bax, Bcl2, and P53 was detected. Masson's trichrome, TUNEL, HE, and PAS staining and transmission electron microscopy were used to detect pathological and ultrastructural changes. Our results showed that rotenone alone and rotenone combined with 3-BrPA significantly increased the risk of ventricular arrhythmias. Rotenone combined with 3-BrPA caused myocardial apoptosis, and rotenone alone and rotenone combined with 3-BrPA caused electrical and structural cardiac remodeling in rats.