Merck
  • Urinary Matrix Metalloproteinase-7 and Prediction of AKI Progression Post Cardiac Surgery.
Urinary Matrix Metalloproteinase-7 and Prediction of AKI Progression Post Cardiac Surgery.
Disease markers (2019-12-13)
Fan Fang, Weihong Luo, Manqiu Yang, Peiliang Yang, Xiaobing Yang
ABSTRACT
Early detection of patients at high risk for progressive acute kidney injury (AKI) after cardiac surgery remains a major challenge. We aim to evaluate the utility of urinary matrix metalloproteinase-7 (uMMP-7) and other reported biomarkers for predicting AKI progression during postoperative hospital stay. We conducted a prospective, multicenter cohort study in 121 adult patients with stage 1 or 2 AKI after cardiac surgery. uMMP-7 and other well-reported biomarkers (uIL-18, uNGAL, and UACR) were measured at time of AKI clinical diagnosis. The primary outcome is the progression of AKI after cardiac surgery, defined as worsening of AKI stage (stage 1 to either stage 2 or stage 3 or from stage 2 to stage 3). A level of uMMP-7 > 7.8 μg/g Cr at time of AKI diagnosis conveyed an 8-fold risk of AKI progression as compared to those with uMMP-7 < 2.7 μg/g after adjusting for clinical risk factors. The performance of uMMP-7 for predicting progressive AKI was good with an AUC of 0.80. The combination of uMMP-7 and IL-18 produces the greatest AUC for predicting progressive AKI. Addition of uMMP-7 to the clinical risk factor model significantly improved risk reclassification for AKI progression. uMMP-7, measured at time of AKI clinical diagnosis, is a novel biomarker for predicting the progression of AKI after cardiac surgery. Adding uMMP-7 to the clinical risk factor model may be used as a noninvasive approach to identify a subpopulation that is at high risk for progressive AKI after cardiac surgery.