- The role of ADGRE5/CD97 in human retinal pigment epithelial cell growth and survival.
The role of ADGRE5/CD97 in human retinal pigment epithelial cell growth and survival.
Cell surface molecules of retinal pigment epithelial (RPE) cells participate in the pathogenesis of retinal diseases. In an attempt to identify cell surface proteins that play a role in RPE cell-cell interactions, we have considered studying the expression, regulation, and signaling of ADGRE5/CD97, an adhesion G protein-coupled receptor family member, based on its known adhesive function in other cell types such as leukocytes. We showed that RPE cells express three isoforms of CD97 and identified inflammation-related cytokines as important mediators regulating CD97 expression. Whereas TNF-α and IFN-γ upregulated CD97, TGF-β decreased CD97 expression. Due to interaction with CD55, ARPE-19 cells firmly adhered to monocytes and T lymphocytes when overexpressing CD97, suggesting a role for CD97 in controlling leukocyte infiltration across the RPE-based blood-retinal barrier. CD97-mediated signaling acted synergistically with PDGF-BB and IFN-γ to regulate cell growth and survival, ensuring a cellular balance under inflammatory conditions. These findings suggest that CD97 on RPE cells serves to control leukocyte activation and trafficking in uveoretinal inflammation while at the same time regulating second messenger-mediated gene expression, cell growth, and survival.