Merck
  • Simultaneous inhibition of neprilysin and activation of ACE2 prevented diabetic cardiomyopathy.
Simultaneous inhibition of neprilysin and activation of ACE2 prevented diabetic cardiomyopathy.
Pharmacological reports : PR (2019-08-31)
Vajir Malek, Nisha Sharma, Anil Bhanudas Gaikwad
ABSTRACT
Neprilysin inhibitors (NEPi) are assisting the renin-angiotensin system (RAS) inhibitors in halting diabetic cardiomyopathy (DCM). Away from conventional tactic, a recent report revealed the renoprotective potential of NEPi and angiotensin-converting enzyme (ACE2) activator combination therapy against diabetic nephropathy. However, this combination so far not evaluated against DCM, thus the present investigation aiming the same. Streptozotocin-induced (55 mg/kg, ip) type 1 diabetic (T1D) male Wistar rats were treated with either monotherapy of thiorphan (0.1 mg/kg/day, po) or diminazene aceturate (5 mg/kg/day, po), or their combination therapy, for four weeks. After hemodynamic measurements, all the rats' heart and plasma were collected for biochemistry, ELISA, histopathology, and immunoblotting. Metabolic perturbations and failing cardiac functions associated with diabetes were markedly attenuated by combination therapy. Besides, unfavourable alterations in RAS and natriuretic peptides system (NPS) were corrected by combination therapy. Interestingly, combination therapy significantly increased plasma and heart cGMP levels compared to T1D and monotherapy receiving rats. Moreover, rats receiving combination therapy exhibited significant inhibition of activated NF-κB, TGF-β and apoptotic signalling, and a notable reduction in cardiac fibrosis when compared to T1D rats. Expressions of posttranslational histone modifications markers; H3K4Me2 and its methyltransferases (SET7/9 and RBBP5) were significantly enhanced in T1D hearts, which were significantly reduced by combination therapy. The NEPi and ACE2 activator combination therapy effectively prevented DCM by normalising RAS and NPS activities, increasing cGMP, inhibiting inflammatory, pro-fibrotic and apoptotic signalling, and reversing H3K4Me2 and its methyl transferases expressions.
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Supelco
Diminazene aceturate, analytical standard