Merck
  • Purification of a 31,000-dalton insulin-like growth factor binding protein from human amniotic fluid. Isolation of two forms with different biologic actions.
Purification of a 31,000-dalton insulin-like growth factor binding protein from human amniotic fluid. Isolation of two forms with different biologic actions.
The Journal of biological chemistry (1988-10-05)
W H Busby, D G Klapper, D R Clemmons
ABSTRACT
Human amniotic fluid has been shown to contain a protein that binds insulin-like growth factor I and II (IGF-I and IGF-II). Partially purified preparations of this protein have been reported to inhibit the biologic actions of the IGFs. In these studies our laboratory has used a modified purification procedure to obtain a homogeneous preparation of this protein as determined by polyacrylamide gel electrophoresis and amino acid sequence analysis. During purification the ion exchange chromatography step resulted in two peaks of material with IGF binding activity termed peaks B and C. Each peak was purified separately to homogeneity. Both peaks were estimated to be 31,000 daltons by polyacrylamide gel electrophoresis and their amino acid compositions were nearly identical. Amino acid sequence analysis showed that both peaks had identical N-terminal sequences through the first 28 residues. Neither protein had detectable carbohydrate side chains and each had a similar affinity for radiolabeled IGF-I (1.7-2.2 x 10(10) liters/mol). In contrast, these two forms had marked differences in bioactivity. Concentrations of peak C material between 2 and 20 ng/ml inhibited IGF-I stimulation of [3H]thymidine incorporation into smooth muscle cell DNA. In contrast, when peak B (100 ng/ml) was incubated with IGF-I there was a 4.4-fold enhancement of stimulation of DNA synthesis. Additionally, pure peak B was shown to adhere to cell surfaces, whereas peak C was not adherent. The non-adherent peak C inhibited IGF-I binding to its receptor and to adherent peak B. We conclude that human amniotic fluid contains two forms of IGF binding protein that have very similar physiochemical characteristics but markedly different biologic actions. Since both have similar if not identical amino acid compositions, N-terminal sequences, and do not contain carbohydrate, we conclude that they differ in some other as yet undefined post-translational modification.