Merck
  • Disseminated tumor cells as selection marker and monitoring tool for secondary adjuvant treatment in early breast cancer. Descriptive results from an intervention study.
Disseminated tumor cells as selection marker and monitoring tool for secondary adjuvant treatment in early breast cancer. Descriptive results from an intervention study.
BMC cancer (2012-12-25)
Marit Synnestvedt, Elin Borgen, Erik Wist, Gro Wiedswang, Kjetil Weyde, Terje Risberg, Christian Kersten, Ingvil Mjaaland, Lise Vindi, Cecilie Schirmer, Jahn Martin Nesland, Bjørn Naume
ABSTRACT
Presence of disseminated tumor cells (DTCs) in bone marrow (BM) after completion of systemic adjuvant treatment predicts reduced survival in breast cancer. The present study explores the use of DTCs to identify adjuvant insufficiently treated patients to be offered secondary adjuvant treatment intervention, and as a surrogate marker for therapy response. A total of 1121 patients with pN1-3 or pT1c/T2G2-3pN0-status were enrolled. All had completed primary surgery and received 6 cycles of anthracycline-containing chemotherapy. BM-aspiration was performed 8-12 weeks after chemotherapy (BM1), followed by a second BM-aspiration 6 months later (BM2). DTC-status was determined by morphological evaluation of immunocytochemically detected cytokeratin-positive cells. If DTCs were present at BM2, docetaxel (100 mg/m², 3qw, 6 courses) was administered, followed by DTC-analysis 1 month (BM3) and 13 months (BM4) after the last docetaxel infusion. Clinical follow-up (FU) is still ongoing. Here, the descriptive data from the study are presented. Of 1085 patients with a reported DTC result at both BM1 and BM2, 94 patients (8.7%) were BM1 positive and 83 (7.6%) were BM2 positive. The concordance between BM1 and BM2 was 86.5%. Both at BM1 and BM2 DTC-status was significantly associated with lobular carcinomas (p = 0.02 and p = 0.03, respectively; chi-square). In addition, DTC-status at BM2 was also associated with pN-status (p = 0.009) and pT-status (p = 0.03). At BM1 28.8% and 12.8% of the DTC-positive patients had ≥2 DTCs and ≥3 DTCs, respectively. At BM2, the corresponding frequencies were 47.0% and 25.3%. Of 72 docetaxel-treated patients analyzed at BM3 and/or BM4, only 15 (20.8%) had persistent DTCs. Of 17 patients with ≥3 DTCs before docetaxel treatment, 12 patients turned negative after treatment (70.6%). The change to DTC-negativity was associated with the presence of ductal carcinoma (p = 0.009). After docetaxel treatment, the majority of patients experienced disappearance of DTCs. As this is not a randomized trial, the results can be due to effects of adjuvant (docetaxel/endocrine/trastuzumab) treatment and/or limitations of the methodology. The clinical significance of these results awaits mature FU data, but indicates a possibility for clinical use of DTC-status as a residual disease-monitoring tool and as a surrogate marker of treatment response. Clin Trials Gov NCT00248703.
Sigma-Aldrich
Anti-Keratin Epithelial Antibody, clone AE3, clone AE3, Chemicon®, from mouse
Sigma-Aldrich
Anti-Cytokeratin Epithelial Antibody, clone AE1, clone AE1, Chemicon®, from mouse