Merck
  • Identification of an unknown glycoprotein from whole cell lysate using conA and mass spectrometry.
Identification of an unknown glycoprotein from whole cell lysate using conA and mass spectrometry.
Biochemistry and molecular biology education : a bimonthly publication of the International Union of Biochemistry and Molecular Biology (2018-07-11)
Jianghong Yan, Shupeng Hu, Xiudan Xu, Dezhang Zhao, Mingjun Wu, Suxin Luo, Yong Xia
ABSTRACT
Medical laboratory technology major was set up to meet rapid development of science and medical research technology in 2013. Students majoring in medical laboratory had learnt a lot of techniques distributed among different specialized courses. But, they did not understand why they had to learn these techniques and how they were applied in a real-world research setting. In a one-month innovation experimental practice described herein, students had learnt to induce, purify and identify an unknown glycoprotein from whole cell lysate using conA-based affinity chromatography and mass spectrometry. Unlike in a traditional cookbook-style experiment, students chose a research subject on their own and did experiment using their selected variables. Over the one-month laboratory periods, students used sterile technique to cultivate cells, induced glycoprotein expression using LPS and IFN-γ, purified glycoprotein from cell lysate using agarose-conA beads, identified a glycoprotein via mass spectrometry, and confirmed the result using western blotting. At end of the practice, students were asked to evaluate their experiences via an anonymous survey. All students declared that this experimental practice was interesting and meaningful to them. The process of completing the project was to apply the learnt techniques to real-world biochemistry research, so they became aware of the importance and significance of techniques. © 2018 by The International Union of Biochemistry and Molecular Biology, 46:373-381, 2018.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), peroxidase conjugate, lyophilized powder
Pricing and availability is not currently available.