Merck
  • Progressive Recruitment of Mesenchymal Progenitors Reveals a Time-Dependent Process of Cell Fate Acquisition in Mouse and Human Nephrogenesis.
Progressive Recruitment of Mesenchymal Progenitors Reveals a Time-Dependent Process of Cell Fate Acquisition in Mouse and Human Nephrogenesis.
Developmental cell (2018-06-06)
Nils O Lindström, Guilherme De Sena Brandine, Tracy Tran, Andrew Ransick, Gio Suh, Jinjin Guo, Albert D Kim, Riana K Parvez, Seth W Ruffins, Elisabeth A Rutledge, Matthew E Thornton, Brendan Grubbs, Jill A McMahon, Andrew D Smith, Andrew P McMahon
ABSTRACT
Mammalian nephrons arise from a limited nephron progenitor pool through a reiterative inductive process extending over days (mouse) or weeks (human) of kidney development. Here, we present evidence that human nephron patterning reflects a time-dependent process of recruitment of mesenchymal progenitors into an epithelial nephron precursor. Progressive recruitment predicted from high-resolution image analysis and three-dimensional reconstruction of human nephrogenesis was confirmed through direct visualization and cell fate analysis of mouse kidney organ cultures. Single-cell RNA sequencing of the human nephrogenic niche provided molecular insights into these early patterning processes and predicted developmental trajectories adopted by nephron progenitor cells in forming segment-specific domains of the human nephron. The temporal-recruitment model for nephron polarity and patterning suggested by direct analysis of human kidney development provides a framework for integrating signaling pathways driving mammalian nephrogenesis.
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer