Skip to Content
Merck
Neomycin trisulfate salt hydrate VETRANAL®, analytical standard

Neomycin sulfate

Neomycin trisulfate salt hydrate VETRANAL®, analytical standard
Synonym(s):
Neomycin trisulfate salt, Neomycin sulphate, (2R,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2-[(2S,3R,4S,5R)-4-[(2R,3R,4R,5S,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol sulfuric acid
Empirical Formula (Hill Notation):
C23H46N6O13 · 3H2SO4
CAS Number:
Molecular Weight:
908.88

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique