N-Nitrosodimethylamine (NDMA) solution
Synonym(s):
N-Methyl-N-nitrosomethanamine, N-Nitrosodimethylamine (NDMA) solution
Empirical Formula (Hill Notation):
C2H6N2O
CAS Number:
Molecular Weight:
74.08
Products
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing