Boron standard solution
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing