Skip to Content
Merck
Ursodeoxycholic acid impurity standard British Pharmacopoeia (BP) Reference Standard

Ursodiol

Ursodeoxycholic acid impurity standard British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Ursodeoxycholic acid, 3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid-3α,7β-diol, 7β-Hydroxylithocholic acid, UDCS
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique