Skip to Content
Merck
Levamisole hydrochloride for system suitability European Pharmacopoeia (EP) Reference Standard

Levamisol hydrochloride

Levamisole hydrochloride for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(−)-Tetramisole hydrochloride, (S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride, (−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo(2,1-b)thiazole, S(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride
Empirical Formula (Hill Notation):
C11H12N2S · HCl
CAS Number:
Molecular Weight:
240.75

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique