Selenomethionine
Synonym(s):
Seleno-L-methionine, (S)-2-Amino-4-(methylseleno)butyric acid
Linear Formula:
CH3SeCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
196.11
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2