Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Economy Pipette Pump III
Products
Product Category
pumps (3)
Manufacturer
Capacity (mL)
Length (in)
Color
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BAF379020002
SP Bel-Art
®
Economy Pipette Pump III,
capacity 2 mL, blue
Expand
View Pricing
BAF379020025
SP Bel-Art
®
Economy Pipette Pump III,
capacity 25 mL, red
Expand
View Pricing
BAF379020010
SP Bel-Art
®
Economy Pipette Pump III,
capacity 10 mL, green
Expand
View Pricing