Skip to Content
Merck
Urea solution suitable for microbiology, 40% in H2O

Urea solution

Urea solution suitable for microbiology, 40% in H2O
Synonym(s):
Carbamide, Carbonyldiamide
Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06

Products

Product Category
Brand
Formula Weight
Gene Alias
pH Value
Physical Form
Product Line
Agency
Quality Segment
Application
Microorganism Suitability
Showing 1-2 of 2
Compare
Product Number
Description
Pricing