Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium permanganate solution
Products
Brand
Supelco (3)
Physical Form
Product Line
Agency
Quality Segment
Reaction Type
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.09121
Potassium permanganate solution,
standardised against sodium thiosulfate c(KMnO
4
) = 0.02 mol/l (0.1 N), Titripur
®
, reag. Ph. Eur.
Expand
View Pricing
1.09122
Potassium permanganate solution,
standardised against oxalate c(KMnO
4
) = 0.02 mol/l (0.1 N), Titripur
®
, reag. USP
Expand
View Pricing
1.60322
Potassium permanganate solution,
c(KMnO
4
) = 0.002 mol; standardised against di-sodium oxalate (CRM), Titripur
®
Expand
View Pricing
4.80160
Potassium permanganate solution,
c(KMnO
4
) = 0.05 mol/l (0.25 N), Titripur
®
Expand
View Pricing