Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly-L-lysine solution
CAS Number:
25988-63-0
Products
Product Category
attachment factors (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Physical Form
Product Line
Quality Segment
Application
Optical Isomer
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P4707
Poly-L-lysine solution,
0.01%, sterile-filtered, BioReagent, suitable for cell culture
Expand
View Pricing
P8920
Poly-L-lysine solution,
0.1 % (w/v) in H
2
O
Expand
View Pricing
P4832
Poly-L-lysine solution,
mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Expand
View Pricing
A-005-M
Poly-L-lysine solution,
0.01%
Expand
View Pricing