Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium carbonate concentrate
Synonym(s):
Sodium carbonate solution
Linear Formula:
Na
2
CO
3
CAS Number:
497-19-8
Molecular Weight:
105.99
Products
Brand
Supelco (2)
Formula Weight
Physical Form
Product Line
Quality Segment
Reaction Type
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
72784
Sodium carbonate concentrate,
Na
2
CO
3
72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Expand
View Pricing
56169
Sodium carbonate concentrate,
0.1 M Na
2
CO
3
in water, eluent concentrate for IC
Expand
View Pricing
1.60310
Sodium carbonate concentrate,
0.05 M Na
2
CO
3
(0.1 N), Titripur
®
Expand
View Pricing