Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
[Pd(OAc)
2
]
3
Synonym(s):
Palladium(II) acetate, Pd(OAc)
2
Linear Formula:
Pd(OCOCH
3
)
2
CAS Number:
3375-31-3
Molecular Weight:
224.51
EC Number:
222-164-4
Products
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
205869
[Pd(OAc)
2
]
3
,
reagent grade, 98%
Expand
View Pricing
520764
[Pd(OAc)
2
]
3
,
≥99.9% trace metals basis
Expand
View Pricing
379875
[Pd(OAc)
2
]
3
,
99.98% trace metals basis
Expand
View Pricing
683124
[Pd(OAc)
2
]
3
,
Umicore, 46.6-49.5% palladium (Pd) basis
Expand
View Pricing