Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MC-Media Pad
®
Products
Product Category
media (5)
Brand
Millipore (5)
Physical Form
Product Line
Agency
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.32359
MC-Media Pad
®
,
for rapid aerobic count
Expand
View Pricing
1.32360
MC-Media Pad
®
,
for yeasts, for molds, Accredited for Food Stuff and Dried Cannabis Flower (THC>0.3%)
Expand
View Pricing
1.32357
MC-Media Pad
®
,
for coliforms, for
Escherichia coli
Expand
View Pricing
1.32356
MC-Media Pad
®
,
for coliforms
Expand
View Pricing
1.32372
MC-Media Pad
®
,
selective for bacteria, Detection of Enterobacteriaceae, pkg of 4 × 25 pieces
Expand
View Pricing