Skip to Content
Merck
vibrant-m

L-Glutamine–Penicillin–Streptomycin solution

vibrant-m

Products

Product Category
Brand
Biological Source
Physical Form
Product Line
Activity Spectrum
Chemical Composition
Quality Segment
Mode of Action
Optical Isomer
Sterilization Method
Technique