Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KDS Stainless Steel Syringe
Products
Product Category
syringes (12)
Feature
Manufacturer
Capacity (mL)
Package Size
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z744197
KDS Stainless Steel Syringe,
capacity 20 mL, joint: Swagelok
®
1/16, pack of 1 ea
Expand
View Pricing
Z744198
KDS Stainless Steel Syringe,
capacity 50 mL, joint: Swagelok
®
1/16, pack of 1 ea
Expand
View Pricing
Z744199
KDS Stainless Steel Syringe,
capacity 100 mL, joint: Swagelok
®
1/16, pack of 1 ea
Expand
View Pricing
Z744200
KDS Stainless Steel Syringe,
capacity 200 mL, joint: Swagelok
®
1/16, pack of 1 ea
Expand
View Pricing
Z744201
KDS Stainless Steel Syringe,
capacity 20 mL, joint: Swagelok
®
1/8, pack of 1 ea
Expand
View Pricing
Z744202
KDS Stainless Steel Syringe,
capacity 50 mL, joint: Swagelok
®
1/8, pack of 1 ea
Expand
View Pricing
Z744203
KDS Stainless Steel Syringe,
capacity 100 mL, joint: Swagelok
®
1/8, pack of 1 ea
Expand
View Pricing
Z744205
KDS Stainless Steel Syringe,
capacity 200 mL, joint: Swagelok
®
1/8, pack of 1 ea
Expand
View Pricing
Z744206
KDS Stainless Steel Syringe,
capacity 20 mL, joint: Swagelok
®
1/4, pack of 1 ea
Expand
View Pricing
Z744207
KDS Stainless Steel Syringe,
capacity 50 mL, joint: Swagelok
®
1/4, pack of 1 ea
Expand
View Pricing
Z744208
KDS Stainless Steel Syringe,
capacity 100 mL, joint: Swagelok
®
1/4, pack of 1 ea
Expand
View Pricing
Z744209
KDS Stainless Steel Syringe,
capacity 200 mL, joint: Swagelok
®
1/4, pack of 1 ea
Expand
View Pricing
Z744210
KDS Stainless Steel Syringe,
Expand
View Pricing