Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gaskets for vertical standard gels, 16.5 cm wide
Products
Product Category
gaskets (3)
Brand
Sigma-Aldrich (3)
Color
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
G6778
Gaskets for vertical standard gels, 16.5 cm wide,
size 1.0 mm × 19 cm, color-coded red
Expand
View Pricing
G3900
Gaskets for vertical standard gels, 16.5 cm wide,
size 1.5 mm × 14.5 cm, color-coded blue
Expand
View Pricing
G3650
Gaskets for vertical standard gels, 16.5 cm wide,
size 1.0 mm × 22 cm, color-coded red
Expand
View Pricing