
Dichloroaniline

Synonym(s):
3,4-Dichloroaniline
Linear Formula:
Cl2C6H3NH2
CAS Number:
Molecular Weight:
162.02
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing