Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CVD graphene oncopper foil, R2R Monolayer large grain
Linear Formula:
C
CAS Number:
1034343-98-0
Products
Brand
Sigma-Aldrich (2)
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
920088
CVD graphene oncopper foil, R2R Monolayer large grain,
A5 size, avg. no. of layers, 1
Expand
View Pricing
920053
CVD graphene oncopper foil, R2R Monolayer large grain,
A3 size, avg. no. of layers, 1
Expand
View Pricing