Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper/Cobalt/Beryllium
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF29858079
Copper/Cobalt/Beryllium,
rod, Cu 97%/Co 2.55/Be 0.5%, 28 mm diameter, length 200 mm
Expand
View Pricing
GF10509678
Copper/Cobalt/Beryllium,
rod, Cu 97%/Co 2.55/Be 0.5%, 28 mm diameter, length 100 mm
Expand
View Pricing
GF44923080
Copper/Cobalt/Beryllium,
rod, Cu 97%/Co 2.55/Be 0.5%, 28 mm diameter, length 500 mm
Expand
View Pricing
GF43738061
Copper/Cobalt/Beryllium,
rod, Cu 97%/Co 2.55/Be 0.5%, 28 mm diameter, length 1000 mm
Expand
View Pricing