
Bacopasaponin C

Synonym(s):
Pseudojujubogenin 3-O-[β-D-glucopyranosyl-(1→3)-[α-L-arabinofuranosyl-(1→2)]-α-L-arabinopyranoside]
Empirical Formula (Hill Notation):
C46H74O17
CAS Number:
Molecular Weight:
899.07
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1