Agar Strips DG-18
Synonym(s):
Agar Strips
Products
Product Category
Brand
Physical Form
Product Line
Compatibility
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing