Skip to Content
Merck
Eosin Y (yellowish) (C.I. 45380) for microscopy Certistain®

Eosin Y (yellowish) (C.I. 45380)

Eosin Y (yellowish) (C.I. 45380) for microscopy Certistain®
Synonym(s):
Eosin Y (yellowish) (C.I. 45380)
Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing