Skip to Content
Merck
(4R,6R)-4-(Ethylamino)-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide-7,7-dioxide monohydrochloride certified reference material, pharmaceutical secondary standard

(4R,6R)-4-(Ethylamino)-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide-7,7-dioxide monohydrochloride

(4R,6R)-4-(Ethylamino)-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide-7,7-dioxide monohydrochloride certified reference material, pharmaceutical secondary standard
Empirical Formula (Hill Notation):
C10H16N2O4S3 · HCl
CAS Number:
Molecular Weight:
360.90

Products

Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade