Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vitamin K
1
Synonym(s):
2-Methyl-3-phytyl-1,4-naphthoquinone, 3-Phytylmenadione, Phylloquinone, Vitamin K
1(20)
Empirical Formula (Hill Notation):
C
31
H
46
O
2
CAS Number:
84-80-0
Molecular Weight:
450.70
EC Number:
201-564-2
Products
Product Category
vitamins (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
V3501
Vitamin K
1
,
viscous liquid
Expand
View Pricing
95271
Vitamin K
1
,
BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Expand
View Pricing