Sodium fluoride
Synonym(s):
Sodium fluoride, Sodium Fluoride
Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing