Hematoxylin
Synonym(s):
Natural Black 1, Hematoxylin cryst. (C.I. 75290)
Empirical Formula (Hill Notation):
C16H14O6
CAS Number:
Molecular Weight:
302.28
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing