Skip to Content
Merck
Iodoacetamide Single use vial of 56 mg

Iodoacetamide

Iodoacetamide Single use vial of 56 mg
Synonym(s):
2-Iodoacetamide
Linear Formula:
ICH2CONH2
CAS Number:
Molecular Weight:
184.96

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Package Size
Reaction Type
Showing 1-6 of 6