Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dodecane
Synonym(s):
n-Dodecane
Linear Formula:
CH
3
(CH
2
)
10
CH
3
CAS Number:
112-40-3
Molecular Weight:
170.33
EC Number:
203-967-9
Products
Product Category
solvents (2)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Anhydrous (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
D221104
Dodecane,
ReagentPlus
®
, ≥99%
Expand
View Pricing
8.20543
Dodecane,
for synthesis
Expand
View Pricing
297879
Dodecane,
anhydrous, ≥99%
Expand
View Pricing