Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Celite
®
S
Synonym(s):
Diatomaceous earth, Diatomaceous silica, Kieselguhr
CAS Number:
61790-53-2
Products
Brand
Sigma-Aldrich (2)
Supelco (1)
Color
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
06858
Celite
®
S,
filter aid, dried, untreated
Expand
View Pricing
1.02051
Celite
®
S,
Absorbent for spilled liquids
Expand
View Pricing
D5509
Celite
®
S,
powder
Expand
View Pricing
525235
Celite
®
S,
meets USP/NF testing specifications
Expand
View Pricing