Calcium chloride
Synonym(s):
Calcium chloride
Linear Formula:
CaCl2
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing