Skip to Content
Merck
Sodium bisulfite anhydrous, free-flowing, Redi-Dri™, ACS reagent

Sodium bisulfite

Sodium bisulfite anhydrous, free-flowing, Redi-Dri™, ACS reagent
Synonym(s):
Sodium hydrogensulfite
Empirical Formula (Hill Notation):
NaHSO3
CAS Number:
Molecular Weight:
104.06

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing