Skip to Content
Merck
All Photos(2)

Key Documents

MAK034

Sigma-Aldrich

Coenzyme A (CoA) Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

functional group

phospholipid

relevant disease(s)

endocrinological disorders, diabetes; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Coenzyme A (CoA) is an essential metabolic cofactor synthesized from cysteine, pantothenate, and ATP (adenosine triphosphate). CoA plays important roles in many metabolic pathways, including the Tricarboxylic Acid cycle, and the synthesis and oxidation of fatty acids. One of the main functions of CoA is the carrying and transfer of acyl groups. Acylated deriviates, for example Acetyl-CoA, are critical intermediates in many metabolic reactions, particularly fatty acid and carbohydrate metabolism. CoA levels can be altered during starvation, and in conditions such as cancer, diabetes, and alcoholism.

Application

Coenzyme A (CoA) Assay Kit has been used to measure CoA levels.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the measurement of Coenzyme A (CoA) in a variety of biological samples including cell and tissue lysates

Principle

The Coenzyme A Assay kit is an easy and convenient assay to measure the CoA level in a variety of biological samples. CoA concentration is determined by an enzymatic assay, which results in a colorimetric (570 nm)/fluorometric (λex = 535 nm/λem = 587 nm) product, proportional to the CoA present. Typical sensitivities of detection for this kit are 0.1-10 nmol of CoA.

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetyl-CoA and the regulation of metabolism: mechanisms and consequences.
Current Opinion in Cell Biology, 33, 125-131 (2015)
ATP-citrate lyase: a key player in cancer metabolism.
Zaidi N, et al.
Cancer Research (2012)
The subcellular localization of acetyl-CoA carboxylase 2.
Abu-Elheiga L, et al.
Proceedings of the National Academy of Sciences of the USA, 97(4), 1444-1449 (2000)
Continuous fatty acid oxidation and reduced fat storage in mice lacking acetyl-CoA carboxylase 2.
Abu-Elheiga L, et al.
Science, 291(5513), 2613-2616 (2001)
Long-Chain Fatty Acyl-CoA Ligase FadD2 Mediates Intrinsic Pyrazinamide Resistance in Mycobacterium tuberculosis.
Rosen B C, et al.
Antimicrobial Agents and Chemotherapy, AAC-02130 (2016)

Articles

Sigma-Aldrich presents an article about how proliferatively active cells require both a source of carbon and of nitrogen for the synthesis of macromolecules. Although a large proportion of tumor cells utilize aerobic glycolysis and shunt metabolites away from mitochondrial oxidative phosphorylation, many tumor cells exhibit increased mitochondrial activity.

Information on fatty acid synthesis and metabolism in cancer cells. Learn how proliferatively active cells require fatty acids for functions such as membrane generation, protein modification, and bioenergetic requirements. These fatty acids are derived either from dietary sources or are synthesized by the cell.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service