Key Documents
DWK73808-18415
Kimble® Closed Top Polypropylene Screw Thread Cap With Welded Ptfe-Faced Silicone Liner
18-415
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
material
PTFE/silicone liner
red polypropylene screw cap (Push fit)
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 73808-18415
fitting
thread
Application
Closure solution for vials and bottles
Features and Benefits
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Closures are ideal for repeated autoclaving
  • Welded liner technology eliminates the possibility of glue contamination
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences