540331
Acetone-d6
99.9 atom % D
Synonym(s):
Hexadeuteroacetone
About This Item
Recommended Products
vapor density
2 (vs air)
vapor pressure
14.39 psi ( 55 °C)
184 mmHg ( 20 °C)
3.67 psi ( 20 °C)
isotopic purity
99.9 atom % D
Assay
99% (CP)
form
liquid
expl. lim.
13.2 %
technique(s)
NMR: suitable
impurities
≤0.0500% water
water
refractive index
n20/D 1.355 (lit.)
bp
55.5 °C (lit.)
mp
−93.8 °C (lit.)
density
0.872 g/mL at 25 °C (lit.)
mass shift
M+6
SMILES string
[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]
InChI
1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3
InChI key
CSCPPACGZOOCGX-WFGJKAKNSA-N
Looking for similar products? Visit Product Comparison Guide
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-2.2 °F - closed cup
Flash Point(C)
-19 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service